Assay Details
| Gene Symbol: Ptgs2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015466.3 | 3 - 4 | 755 | 75 | ||
| GenBank mRNA | AK083274.1 | - | 3 - 4 | 699 | 75 | |
| AK144829.1 | - | 3 - 4 | 648 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
phospholipase A2, group IVA (cytosolic, calcium-dependent) |
Pla2g4a |
Pla2g4, cPLA2, cPLA2-alpha, cPLA2alpha |
-123.3kb | Sense | Chr. 1 - 149829618 - 149961284 on NCBI Build GRCm38 |
| Downstream | prostaglandin-endoperoxide synthase 2 | Ptgs2 |
COX2, Cox-2, PGHS-2, PHS-2, Pghs2, TIS10 |
12.8kb | Anti-sense | Chr. 1 - 150100124 - 150108017 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Ptgs2os
Genomic Map