Assay Details
| Gene Symbol: Mir99ahg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015543.2 | 2 - 3 | 411 | 108 | ||
| GenBank mRNA | AK083771.1 | - | 2 - 3 | 412 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin specific peptidase 25 | Usp25 | -243.9kb | Sense | Chr. 16 - 77013789 - 77116783 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 31322 |
Gm31322 | 129.9kb | Sense | Chr. 16 - 77589379 - 77790662 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Mir99ahg
Genomic Map