Assay Details
| Gene Symbol: 9330111N05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015587.2 | 1 - 2 | 315 | 82 | ||
| GenBank mRNA | AK165883.1 | - | 1 - 2 | 316 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 4211 | Gm4211 | -2.6kb | Anti-sense | Chr. 13 - 80957664 - 80961434 on NCBI Build GRCm38 | |
| Downstream | adhesion G protein-coupled receptor V1 | Adgrv1 |
Frings, Gpr98, Mass1, Mgr1, VLGR1 |
15.2kb | Anti-sense | Chr. 13 - 81095068 - 81633197 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

9330111N05Rik
Genomic Map