Assay Details
| Gene Symbol: 4932441J04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015588.2 | 1 - 2 | 77 | 102 | ||
| GenBank mRNA | AK039574.1 | - | 1 - 2 | 78 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protocadherin 7 | Pcdh7 | Anti-sense | Chr. 5 - 57716456 - 58133235 on NCBI Build GRCm38 | ||
| Downstream | RIKEN cDNA 4930459L07 gene | 4930459L07Rik | 725.3kb | Sense | Chr. 5 - 58443239 - 58452998 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4932441J04Rik
Genomic Map