Assay Details
| Gene Symbol: 4932441J04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015588.2 | 4 - 5 | 396 | 146 | ||
| GenBank mRNA | AK039574.1 | - | 4 - 5 | 397 | 146 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 10440 | Gm10440 |
D130065D13, ENSMUSG00000072936 |
-3.2mb | Anti-sense | Chr. 5 - 54349992 - 54358542 on NCBI Build GRCm38 |
| Downstream | protocadherin 7 | Pcdh7 | 115.0kb | Anti-sense | Chr. 5 - 57716456 - 58133235 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4932441J04Rik
Genomic Map