Assay Details
| Gene Symbol: AF357355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028433.2 | 1 | 33 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | snoRNA DQ267101 | DQ267101 | -3.1kb | Sense | Chr. 12 - 109651424 - 109651495 on NCBI Build GRCm38 | |
| Downstream | snoRNA DQ267102 | DQ267102 | 1.6kb | Sense | Chr. 12 - 109656345 - 109656415 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

AF357355
Genomic Map