Assay Details
| Gene Symbol: Snord92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028556.2 | 1 | 45 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WD repeat domain 43 | Wdr43 |
2610318G08Rik, AU020887, AV024208, Wrd43, mKIAA0007 |
Sense | Chr. 17 - 71615900 - 71659031 on NCBI Build GRCm38 | |
| Downstream |
small nucleolar RNA, C/D box 53 |
Snord53 | MBII-35 | 9.5kb | Sense | Chr. 17 - 71640889 - 71640956 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snord92
Genomic Map