Assay Details
| Gene Symbol: Mir187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029573.1 | 1 | 31 | 61 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 38410 |
Gm38410 | 9530053H22 | -84.5kb | Anti-sense | Chr. 18 - 24339366 - 24344630 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 2700062C07 gene | 2700062C07Rik |
AI195775, C87515 |
41.7kb | Anti-sense | Chr. 18 - 24470871 - 24477767 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir187
Genomic Map