Assay Details
| Gene Symbol: Mir302a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029653.1 | 1 | 29 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 302c | Mir302c |
Mirn302c, mir-302c |
-66.0 | Sense | Chr. 3 - 127545363 - 127545430 on NCBI Build GRCm38 |
| Downstream | microRNA 302d | Mir302d |
Mirn302d, mir-302d |
60.0 | Sense | Chr. 3 - 127545624 - 127545689 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir302a
Genomic Map