Assay Details
| Gene Symbol: Mir328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029761.1 | 1 | 43 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | engulfment and cell motility 3 | Elmo3 |
9930107J06, BC058752, CED-12, Ced12 |
Anti-sense | Chr. 8 - 105305399 - 105310623 on NCBI Build GRCm38 | |
| Downstream | leucine rich repeat containing 29 | Lrrc29 |
A630024J02Rik, FBL9, FBXL9 |
3.9kb | Sense | Chr. 8 - 105312340 - 105326331 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir328
Genomic Map