Assay Details
| Gene Symbol: Mir302d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030405.1 | 1 | 30 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 302a | Mir302a |
Mirn302, Mirn302a, mir-302a, mmu-mir-302 |
-60.0 | Sense | Chr. 3 - 127545496 - 127545564 on NCBI Build GRCm38 |
| Downstream | microRNA 367 | Mir367 |
Mirn367, mir-367, mmu-mir-367 |
44.0 | Sense | Chr. 3 - 127545733 - 127545807 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir302d
Genomic Map