Assay Details
| Gene Symbol: Mir2137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_035517.1 | 1 | 35 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 185 | Zfp185 |
P1, Znf185 |
Sense | Chr. X - 72987253 - 73031543 on NCBI Build GRCm38 | |
| Downstream | paraneoplastic antigen family 5 | Pnma5 | mKIAA1934 | 41.8kb | Anti-sense | Chr. X - 73033981 - 73040203 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir2137
Genomic Map