Assay Details
| Gene Symbol: Asmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199212.1 | NP_001186141.1 | 3 - 4 | 444 | 77 | |
| GenBank mRNA | AB512670.1 | - | 3 - 4 | 419 | 77 | |
| AB512671.1 | - | 3 - 4 | 444 | 77 | ||
| AB512672.1 | - | 3 - 4 | 396 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Asmt
Genomic Map