Assay Details
| Gene Symbol: Asmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199212.1 | NP_001186141.1 | 7 - 8 | 914 | 73 | |
| GenBank mRNA | AB512670.1 | - | 7 - 8 | 889 | 73 | |
| AB512671.1 | - | 7 - 8 | 914 | 73 | ||
| AB512672.1 | - | 7 - 8 | 866 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Asmt
Genomic Map