Assay Details
| Gene Symbol: Mir5104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_039563.1 | 1 | 48 | 86 | ||
| GenBank mRNA | JF446403.1 | - | 1 | 48 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger CCCH type containing 12D | Zc3h12d |
D730019B10Rik, TFL |
Sense | Chr. 10 - 7832323 - 7870397 on NCBI Build GRCm38 | |
| Downstream | TGF-beta activated kinase 1/MAP3K7 binding protein 2 | Tab2 |
1110030N06Rik, A530078N03Rik, Map3k7ip2, mKIAA0733 |
69.2kb | Anti-sense | Chr. 10 - 7905648 - 7956225 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir5104
Genomic Map