Assay Details
Gene Symbol: Mir5110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_039570.1 | 1 | 44 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | breast carcinoma amplified sequence 3 | Bcas3 |
1500019F07Rik, 2610028P08Rik, AU021018, BC028339, K20D4, rudhira |
Anti-sense | Chr. 11 - 85353116 - 85826058 on NCBI Build GRCm38 | |
Downstream | breast carcinoma amplified sequence 3 opposite strand 2 | Bcas3os2 | 9530048J24Rik | 6.1kb | Sense | Chr. 11 - 85766833 - 85775680 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
