Assay Details
| Gene Symbol: Mir5120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_039580.1 | 1 | 38 | 51 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | paired box 5 | Pax5 |
BSAP, EBB-1, KLP, Pax-5 |
Sense | Chr. 4 - 44526269 - 44710481 on NCBI Build GRCm38 | |
| Downstream | uncharacterized LOC102632357 | LOC102632357 | 52.8kb | Anti-sense | Chr. 4 - 44660394 - 44683684 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir5120
Genomic Map