Assay Details
Gene Symbol: Mir5125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_039586.1 | 1 | 36 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | FLYWCH family member 2 | Flywch2 |
2810417J12Rik, 3010014L17Rik, AW548203 |
-37.2kb | Anti-sense | Chr. 17 - 23776918 - 23786077 on NCBI Build GRCm38 |
Downstream |
transcription elongation factor B (SIII), polypeptide 2 |
Tceb2 | 0610040H15Rik | 1.4kb | Anti-sense | Chr. 17 - 23824740 - 23829109 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
