Assay Details
| Gene Symbol: 1700029N11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045489.1 | 1 - 2 | 130 | 127 | ||
| GenBank mRNA | AK006512.1 | - | 1 - 2 | 131 | 127 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | 
						predicted gene, 29676  | 
					Gm29676 | -179.6kb | Anti-sense | Chr. 13 - 44250092 - 44260123 on NCBI Build GRCm38 | |
| Downstream | 
						jumonji, AT rich interactive domain 2  | 
					Jarid2 | 
					Jmj, jumonji  | 
					273.9kb | Sense | Chr. 13 - 44729181 - 44921643 on NCBI Build GRCm38 | 
More Information
Updated Mapping Information
Set Membership: | 
		   
					
						
							 | 
				

1700029N11Rik
Genomic Map