Assay Details
Gene Symbol: 1700029N11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045489.1 | 2 - 3 | 217 | 68 | ||
GenBank mRNA | AK006512.1 | - | 2 - 3 | 218 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 29676 |
Gm29676 | -195.1kb | Anti-sense | Chr. 13 - 44250092 - 44260123 on NCBI Build GRCm38 | |
Downstream |
jumonji, AT rich interactive domain 2 |
Jarid2 |
Jmj, jumonji |
272.7kb | Sense | Chr. 13 - 44729181 - 44921643 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 401625 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720257 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652145 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 531215 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361867 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 835033 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 53983 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07857492 | - | - |
Whole Mouse Genome G4122A | A_52_P962354 | AK006512.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124954 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11090887 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996062 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996038 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996043 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996080 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996040 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996099 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10864293 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996054 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996136 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996167 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996120 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996090 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10945559 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996079 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10906731 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996218 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996097 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10528928 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996056 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10831392 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996071 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996069 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996117 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996061 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10464144 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996220 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996224 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10906732 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996132 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996065 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996112 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996199 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996100 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996121 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996180 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10652609 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996197 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996053 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996130 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996238 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996204 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10596606 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996107 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996036 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11091973 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996234 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10511864 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996181 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10873174 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10520445 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10464149 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10873170 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10833732 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996084 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996207 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996191 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996125 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996193 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996203 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996163 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996206 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996128 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996088 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996032 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996113 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996111 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996174 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996212 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996233 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996104 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10576710 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996066 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10511863 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10873179 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996087 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996033 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996239 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996187 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996188 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996045 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996182 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996184 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996105 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11033833 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10705956 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996048 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996072 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996222 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996089 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996172 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996122 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996227 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10864288 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996067 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996219 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996209 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996092 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996102 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996126 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996231 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996073 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996098 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10907978 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11043440 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996044 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996168 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996076 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996237 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10508518 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996114 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996095 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10888974 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996127 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996063 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996030 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996124 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996057 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996034 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28996119 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR1300003100.mm.1 | NR_045489.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300003100.mm.1 | NR_045489.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300003100.mm.1 | TC1300000512.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300003098.mm.1 | NR_045489.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300003098.mm.1 | NR_045489.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300003098.mm.1 | TC1300000512.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 353805 | - | - |
