Assay Details
| Gene Symbol: Lmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045520.1 | 1 - 2 | 203 | 82 | ||
| GenBank mRNA | AK041659.1 | - | 1 - 2 | 191 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 2810468N07 gene | 2810468N07Rik | -4.1kb | Sense | Chr. 17 - 25570811 - 25575043 on NCBI Build GRCm38 | |
| Downstream |
guanine nucleotide binding protein (G protein), gamma 13 |
Gng13 |
1500031D04Rik, AB030194, Ggamma13 |
104.7kb | Sense | Chr. 17 - 25717172 - 25719102 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041659.1 transcripts.
Set Membership: |
|

Lmf1
Genomic Map