Assay Details
| Gene Symbol: Sh2d3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045535.1 | 3 - 4 | 648 | 94 | ||
| GenBank mRNA | AK172072.1 | - | 3 - 4 | 569 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3960 | Mir3960 | mmu-mir-3960 | -24.5kb | Anti-sense | Chr. 2 - 32712900 - 32712972 on NCBI Build GRCm38 |
| Downstream | tetratricopeptide repeat domain 16 | Ttc16 | 1200002K10Rik | 15.4kb | Anti-sense | Chr. 2 - 32757026 - 32775663 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Sh2d3c
Genomic Map