Assay Details
| Gene Symbol: Slc35c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045544.1 | 1 - 2 | 53 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cadherin 22 | Cdh22 | -48.6kb | Sense | Chr. 2 - 165111501 - 165234879 on NCBI Build GRCm38 | |
| Downstream | engulfment and cell motility 2 | Elmo2 |
1190002F24Rik, CED-12 |
193.0 | Sense | Chr. 2 - 165288031 - 165326479 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Slc35c2
Genomic Map