Assay Details
| Gene Symbol: 1110017D15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045549.2 | 4 - 5 | 633 | 119 | ||
| NR_045551.2 | 1 - 2 | 240 | 119 | |||
| GenBank mRNA | BC049629.1 | - | 1 - 2 | 230 | 119 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | expressed sequence AI464131 | AI464131 |
AA408153, AI604836, Gm762, Kiaa1161, NET37 |
-4.0kb | Sense | Chr. 4 - 41491847 - 41503136 on NCBI Build GRCm38 |
| Downstream |
family with sequence similarity 219, member A, opposite strand |
Fam219aos | 1700066J24Rik | 9.7kb | Anti-sense | Chr. 4 - 41517437 - 41518929 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1110017D15Rik
Genomic Map