Assay Details
| Gene Symbol: Fut8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045554.1 | 1 - 2 | 516 | 68 | ||
| GenBank mRNA | AK133547.1 | - | 1 - 2 | 399 | 68 | |
| AK134537.1 | - | 1 - 2 | 190 | 68 | ||
| AK157989.1 | - | 1 - 2 | 443 | 68 | ||
| AK163494.1 | - | 1 - 2 | 213 | 68 | ||
| CJ070536.1 | - | 1 - 2 | 516 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Max protein | Max |
AA960152, AI875693, bHLHd4, bHLHd5, bHLHd6, bHLHd7, bHLHd8 |
-276.4kb | Anti-sense | Chr. 12 - 76937269 - 76962335 on NCBI Build GRCm38 |
| Downstream | gephyrin | Gphn |
5730552E08Rik, AI662856, BC027112, C230040D23, GPH, GPHRYN, geph |
946.7kb | Sense | Chr. 12 - 78224153 - 78684772 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK134537.1 transcripts.
Set Membership: |
|

Fut8
Genomic Map