Assay Details
| Gene Symbol: Plk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045576.1 | 4 - 5 | 640 | 98 | ||
| GenBank mRNA | AK164465.1 | - | 4 - 5 | 550 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | heat shock protein 4 like | Hspa4l |
94kDa, AI461691, APG-1, Osp94 |
-5.9kb | Sense | Chr. 3 - 40744355 - 40796103 on NCBI Build GRCm38 |
| Downstream | major facilitator superfamily domain containing 8 | Mfsd8 |
2810423E13Rik, AI836898, AV142426, Cln7 |
12.2kb | Anti-sense | Chr. 3 - 40818112 - 40846856 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Plk4
Genomic Map