Assay Details
| Gene Symbol: Gm4961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045694.2 | 1 - 2 | 107 | 124 | ||
| GenBank mRNA | AK139799.1 | - | 1 - 2 | 94 | 124 | |
| CF173951.1 | - | 1 - 2 | 107 | 124 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | thymidylate synthase | Tyms | Ts | -19.3kb | Anti-sense | Chr. 5 - 30058200 - 30073625 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 3110082J24 gene | 3110082J24Rik | ENSMUSG00000073106 | 5.9kb | Anti-sense | Chr. 5 - 30103584 - 30106086 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0500001289.rn
Set Membership: |
|

Gm4961
Genomic Map