Assay Details
| Gene Symbol: Gm19299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045748.1 | 2 - 3 | 275 | 100 | ||
| GenBank mRNA | AK136121.1 | - | 2 - 3 | 277 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
tropomyosin 1, alpha |
Tpm1 |
AA986836, AI854628, TM2, Tm3, Tmpa, Tpm-1, alpha-TM |
-107.6kb | Anti-sense | Chr. 9 - 67022590 - 67049401 on NCBI Build GRCm38 |
| Downstream | talin 2 | Tln2 |
5730421P04Rik, AI507121, AI787438, AL118320, mKIAA0320 |
53.3kb | Anti-sense | Chr. 9 - 67217085 - 67634036 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 762302 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187126 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739961 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 585217 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 163741 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412402 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 867163 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59334 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 270638 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777522 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 820287 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 122937 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172775 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01959367 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172775 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01959352 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07482468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07483674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07539927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07921629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07554713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07723284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07496498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07590172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07713992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08161661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08161670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08161662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07713977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08117311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07553225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07744609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07865295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07713990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07749890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08117305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26597261 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR0900006811.mm.1 | NR_045748.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006811.mm.1 | NR_045748.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006811.mm.1 | TC0900000878.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900006810.mm.1 | NR_045748.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006810.mm.1 | NR_045748.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006810.mm.1 | TC0900000878.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900006812.mm.1 | NR_045748.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006812.mm.1 | NR_045748.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006812.mm.1 | TC0900000878.mm.1 | MTA TC |

Gm19299
Genomic Map