Assay Details
| Gene Symbol: 4921511I17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045829.1 | 2 - 3 | 168 | 88 | ||
| GenBank mRNA | AK014875.1 | - | 2 - 3 | 169 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
tubulin, beta 2a, pseudogene 2 |
Tubb2a-ps2 | C79445 | -490.1kb | Sense | Chr. 12 - 11882196 - 11882899 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 34637 |
Gm34637 | 276.6kb | Sense | Chr. 12 - 12667732 - 12669963 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4921511I17Rik
Genomic Map