Assay Details
| Gene Symbol: Gm10532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045879.1 | 1 - 2 | 102 | 74 | ||
| GenBank mRNA | AK133424.1 | - | 1 - 2 | 103 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CBP80/20-dependent translation initiation factor | Ctif | Gm672 | Anti-sense | Chr. 18 - 75431205 - 75697696 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 1700034B16 gene | 1700034B16Rik | 262.1kb | Sense | Chr. 18 - 75781031 - 75782301 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK133424.1 transcripts.
Set Membership: |
|

Gm10532
Genomic Map