Assay Details
| Gene Symbol: Gm15612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045880.1 | 1 - 2 | 245 | 78 | ||
| GenBank mRNA | AK133241.1 | - | 1 - 2 | 246 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ankyrin repeat and BTB (POZ) domain containing 1 | Abtb1 |
AI847549, BC003234, BPOZ, EF1ABP |
-468.0 | Anti-sense | Chr. 6 - 88835914 - 88842386 on NCBI Build GRCm38 |
| Downstream | minichromosome maintenance deficient 2 mitotin (S. cerevisiae) | Mcm2 |
AA959861, AW476101, BM28, CDCL1, Mcmd2, mKIAA0030 |
39.4kb | Anti-sense | Chr. 6 - 88883474 - 88898780 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK133241.1 transcripts.
Set Membership: |
|

Gm15612
Genomic Map