Assay Details
| Gene Symbol: Gm10556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045881.1 | 6 - 7 | 704 | 62 | ||
| GenBank mRNA | AK138579.1 | - | 6 - 7 | 706 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 9430020K01 gene | 9430020K01Rik |
1110056P05Rik, Gm328, Jcad, mKIAA1462 |
-162.0kb | Sense | Chr. 18 - 4590790 - 4682869 on NCBI Build GRCm38 |
| Downstream | supervillin | Svil |
AU024053, B430302E16Rik |
72.3kb | Sense | Chr. 18 - 4920236 - 5119293 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK138579.1 transcripts.
Set Membership: |
|

Gm10556
Genomic Map