Assay Details
| Gene Symbol: Gm10578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045885.1 | 2 - 3 | 244 | 77 | ||
| GenBank mRNA | AK133282.1 | - | 2 - 3 | 245 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | O-6-methylguanine-DNA methyltransferase | Mgmt |
AGT, AI267024, Agat |
-108.7kb | Sense | Chr. 7 - 136894468 - 137128193 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 9430038I01 gene | 9430038I01Rik | 137.1kb | Anti-sense | Chr. 7 - 137375569 - 137411900 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm10578
Genomic Map