Assay Details
| Gene Symbol: Gm10538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045892.1 | 2 - 3 | 1166 | 97 | ||
| GenBank mRNA | AK133041.1 | - | 2 - 3 | 1167 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neurofascin | Nfasc |
AA387016, D430023G06Rik, NF, mKIAA0756 |
Anti-sense | Chr. 1 - 132564690 - 132741834 on NCBI Build GRCm38 | |
| Downstream |
leucine rich repeat protein 2, neuronal |
Lrrn2 |
5730406J09Rik, NLRR-2 |
147.5kb | Sense | Chr. 1 - 132880355 - 132940005 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK133041.1 transcripts.
Set Membership: |
|

Gm10538
Genomic Map