Assay Details
| Gene Symbol: AU016765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045899.1 | 2 - 3 | 366 | 79 | ||
| GenBank mRNA | AK139806.1 | - | 2 - 3 | 367 | 79 | |
| BG083599.1 | - | 2 - 3 | 79 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6420 | Mir6420 |
Gm28007, mmu-mir-6420 |
-75.2kb | Anti-sense | Chr. 17 - 64440806 - 64440907 on NCBI Build GRCm38 |
| Downstream |
mannosidase 2, alpha 1 |
Man2a1 |
Mana-2, Mana2, Map-2 |
81.6kb | Anti-sense | Chr. 17 - 64601649 - 64755110 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

AU016765
Genomic Map