Assay Details
| Gene Symbol: AU016765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045899.1 | 3 - 4 | 589 | 119 | ||
| GenBank mRNA | AK139806.1 | - | 3 - 4 | 590 | 119 | |
| BG083599.1 | - | 1 - 2 | 119 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6420 | Mir6420 |
Gm28007, mmu-mir-6420 |
-73.2kb | Anti-sense | Chr. 17 - 64440806 - 64440907 on NCBI Build GRCm38 |
| Downstream |
mannosidase 2, alpha 1 |
Man2a1 |
Mana-2, Mana2, Map-2 |
85.3kb | Anti-sense | Chr. 17 - 64601649 - 64755110 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118124 | AK139806.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118124 | AU016765.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102636 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118124 | AK139806.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118124 | AU016765.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08828272 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102636 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08828290 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13422225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13061128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13407932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13589381 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13422236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295898 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13407832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13407940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295873 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13407911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13444552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13204009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13407922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13121092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13164725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13407938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296156 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13061124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13422246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13422232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13407923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13164729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296098 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13121110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13407826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13422231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13121094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13422252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13422226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13543112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13206665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31295924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13407928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13263258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13061122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31296182 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR1700022327.mm.1 | NR_045899.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700022327.mm.1 | NR_045899.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700022327.mm.1 | TC1700002404.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700022324.mm.1 | NR_045899.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700022324.mm.1 | NR_045899.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700022324.mm.1 | TC1700002404.mm.1 | MTA TC |

AU016765
Genomic Map