Assay Details
| Gene Symbol: Gm15008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045917.1 | 2 - 3 | 147 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | septin 6 | Sept6 |
2810035H17Rik, C920001C06Rik, Sep6, mKIAA0128 |
Anti-sense | Chr. X - 36911272 - 36989695 on NCBI Build GRCm38 | |
| Downstream | sosondowah ankyrin repeat domain family member D | Sowahd |
A630014H24Rik, Ankrd58 |
137.2kb | Sense | Chr. X - 37048845 - 37050418 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm15008
Genomic Map