Assay Details
| Gene Symbol: Slfn5os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045932.1 | 1 - 2 | 74 | 105 | ||
| GenBank mRNA | AK005634.1 | - | 1 - 2 | 51 | 105 | |
| BY705829.1 | - | 1 - 2 | 51 | 105 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | unc-45 homolog B (C. elegans) | Unc45b |
AA445617, Cmya4, D230041A13Rik, Unc45 |
-45.0 | Anti-sense | Chr. 11 - 82911253 - 82943406 on NCBI Build GRCm38 |
| Downstream | schlafen 9 | Slfn9 | 9830137M10Rik | 17.7kb | Sense | Chr. 11 - 82978382 - 82992233 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005634.1 transcripts.
Set Membership: |
|

Slfn5os
Genomic Map