Assay Details
| Gene Symbol: Gm10445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046063.1 | 1 - 2 | 273 | 77 | ||
| GenBank mRNA | AK144571.1 | - | 1 - 2 | 274 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dysferlin | Dysf |
2310004N10Rik, AI604795, D6Pas3 |
-210.5kb | Anti-sense | Chr. 6 - 84008316 - 84211060 on NCBI Build GRCm38 |
| Downstream |
cytochrome P450, family 26, subfamily b, polypeptide 1 |
Cyp26b1 |
CP26, P450RAI-2 |
147.6kb | Sense | Chr. 6 - 84571414 - 84593908 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm10445
Genomic Map