Assay Details
| Gene Symbol: Gm15850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046169.1 | 1 - 2 | 166 | 167 | ||
| NR_046170.1 | 1 - 2 | 166 | 167 | |||
| GenBank mRNA | BC107310.1 | - | 1 - 2 | 154 | 167 | |
| BC114580.1 | - | 1 - 2 | 154 | 167 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
calcium channel, voltage-dependent, L type, alpha 1S subunit |
Cacna1s |
AW493108, Cav1.1, Cchl1a3, fmd, mdg, sj |
-9.6kb | Anti-sense | Chr. 1 - 136052805 - 136119822 on NCBI Build GRCm38 |
| Downstream | kinesin family member 21B | Kif21b |
2610511N21Rik, mKIAA0449 |
191.0 | Anti-sense | Chr. 1 - 136131374 - 136178014 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm15850
Genomic Map