Assay Details
| Gene Symbol: Rubie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046459.1 | 1 - 2 | 171 | 60 | ||
| GenBank mRNA | BY232928.1 | - | 1 - 2 | 167 | 60 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA E130120K24 gene | E130120K24Rik | -2.7kb | Anti-sense | Chr. 14 - 46546345 - 46568344 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4933425B07 gene | 4933425B07Rik | 27.0kb | Sense | Chr. 14 - 46602806 - 46637423 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Rubie
Genomic Map