Assay Details
| Gene Symbol: Rubie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046459.1 | 4 - 5 | 990 | 94 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA E130120K24 gene | E130120K24Rik | Anti-sense | Chr. 14 - 46546345 - 46568344 on NCBI Build GRCm38 | ||
| Downstream | RIKEN cDNA 4933425B07 gene | 4933425B07Rik | 33.0kb | Sense | Chr. 14 - 46602806 - 46637423 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Rubie
Genomic Map