Assay Details
| Gene Symbol: Flcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271356.1 | NP_001258285.1 | 10 - 11 | 1780 | 85 | |
| NM_001271357.1 | NP_001258286.1 | 10 - 11 | 1523 | 85 | ||
| NM_146018.2 | NP_666130.1 | 10 - 11 | 1820 | 85 | ||
| NR_073164.1 | 10 - 11 | 2091 | 85 | |||
| GenBank mRNA | AK039106.1 | - | 10 - 11 | 1607 | 85 | |
| AK080933.1 | - | 10 - 11 | 1605 | 85 | ||
| AK136071.1 | - | 10 - 11 | 1511 | 85 | ||
| AK148216.1 | - | 10 - 11 | 1574 | 85 | ||
| AK154040.1 | - | 10 - 11 | 1802 | 85 | ||
| AK161460.1 | - | 10 - 11 | 1863 | 85 | ||
| AK172245.1 | - | 10 - 11 | 1578 | 85 | ||
| AK177301.1 | - | 1 - 2 | 102 | 85 | ||
| AK213109.1 | - | 3 - 4 | 85 | |||
| BC025820.1 | - | 10 - 11 | 1554 | 5099493 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Flcn
Genomic Map