Assay Details
| Gene Symbol: Flcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271356.1 | NP_001258285.1 | 4 - 5 | 877 | 81 | |
| NM_001271357.1 | NP_001258286.1 | 4 - 5 | 620 | 81 | ||
| NM_146018.2 | NP_666130.1 | 4 - 5 | 917 | 81 | ||
| NR_073164.1 | 4 - 5 | 877 | 81 | |||
| GenBank mRNA | AK039106.1 | - | 4 - 5 | 704 | 81 | |
| AK080933.1 | - | 4 - 5 | 702 | 81 | ||
| AK136071.1 | - | 4 - 5 | 608 | 81 | ||
| AK148216.1 | - | 4 - 5 | 671 | 81 | ||
| AK154040.1 | - | 4 - 5 | 899 | 81 | ||
| AK161460.1 | - | 4 - 5 | 649 | 81 | ||
| AK172245.1 | - | 4 - 5 | 676 | 81 | ||
| AK182652.1 | - | 1 - 2 | 81 | |||
| BC025820.1 | - | 4 - 5 | 651 | 5099493 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Flcn
Genomic Map