Assay Details
| Gene Symbol: Flcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271356.1 | NP_001258285.1 | 5 - 6 | 1099 | 75 | |
| NM_001271357.1 | NP_001258286.1 | 5 - 6 | 842 | 75 | ||
| NM_146018.2 | NP_666130.1 | 5 - 6 | 1139 | 75 | ||
| NR_073164.1 | 5 - 6 | 1099 | 75 | |||
| GenBank mRNA | AK039106.1 | - | 5 - 6 | 926 | 75 | |
| AK080933.1 | - | 5 - 6 | 924 | 75 | ||
| AK136071.1 | - | 5 - 6 | 830 | 75 | ||
| AK148216.1 | - | 5 - 6 | 893 | 75 | ||
| AK154040.1 | - | 5 - 6 | 1121 | 75 | ||
| AK161460.1 | - | 5 - 6 | 871 | 75 | ||
| AK172245.1 | - | 5 - 6 | 898 | 75 | ||
| BC025820.1 | - | 5 - 6 | 873 | 5099493 | 75 | |
More Information
Set Membership: |
|

Flcn
Genomic Map