Assay Details
| Gene Symbol: Clip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271483.1 | NP_001258412.1 | 7 - 8 | 1037 | 71 | |
| NM_001271484.1 | NP_001258413.1 | 7 - 8 | 1122 | 71 | ||
| NM_030179.3 | NP_084455.2 | 7 - 8 | 1025 | 71 | ||
| NM_175378.1 | NP_780587.1 | 7 - 8 | 1122 | 71 | ||
| NR_073185.1 | 7 - 8 | 1122 | 71 | |||
| GenBank mRNA | AK017914.1 | - | 4 - 5 | 613 | 71 | |
| AK019516.1 | - | 7 - 8 | 1038 | 71 | ||
| AK029962.1 | - | 7 - 8 | 1123 | 71 | ||
| AK075758.1 | - | 7 - 8 | 1113 | 71 | ||
| AK161301.1 | - | 7 - 8 | 1112 | 71 | ||
| BC035226.1 | - | 7 - 8 | 1025 | 4920788 | 71 | |
More Information
Set Membership: |
|

Clip4
Genomic Map