Assay Details
| Gene Symbol: Snora64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002897.1 | 1 | 48 | 68 | ||
| GenBank mRNA | AF357417.1 | - | 1 | 48 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein S2 | Rps2 |
Llrep3, S4 |
Sense | Chr. 17 - 24718121 - 24721927 on NCBI Build GRCm38 | |
| Downstream |
NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10 |
Ndufb10 |
0610011B04Rik, 22kDa, PDSW |
1.2kb | Anti-sense | Chr. 17 - 24722067 - 24724388 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snora64
Genomic Map