Assay Details
| Gene Symbol: 1700120C14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045627.2 | 1 | 173 | 89 | ||
| GenBank mRNA | AK007210.1 | - | 1 | 174 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microspherule protein 1 | Mcrs1 |
C78274, ICP22BP, MSP58, P78 |
Anti-sense | Chr. 15 - 99242817 - 99251961 on NCBI Build GRCm38 | |
| Downstream |
family with sequence similarity 186, member B |
Fam186b | EG545136 | 19.0kb | Anti-sense | Chr. 15 - 99271018 - 99295891 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700120C14Rik
Genomic Map