Assay Details
| Gene Symbol: 1700020G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045979.1 | 3 | 502 | 116 | ||
| GenBank mRNA | AK076938.1 | - | 3 | 503 | 116 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | E2F transcription factor 7 | E2f7 |
A630014C11Rik, D10Ertd739e |
-13.8kb | Anti-sense | Chr. 10 - 110745427 - 110787384 on NCBI Build GRCm38 |
| Downstream | cysteine and glycine-rich protein 2 | Csrp2 |
AW551867, Crp2, SmLim |
118.7kb | Anti-sense | Chr. 10 - 110920176 - 110939514 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 403618 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 116200 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 590197 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 800750 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783446 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 769878 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | AF276950.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | AF276951.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | AF276952.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | AF454943.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | AF454944.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | AK017487.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | AK029532.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | AK029747.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | AK049516.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | AK050488.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | AK157813.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | AK167724.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | BC019456.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | BC024449.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | BC038874.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | BC057869.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767390 | NM_027444.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218975 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439121 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 280072 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 258864 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 708608 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 696743 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715414 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335953 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45918 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 65850 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204572 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278990 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447811 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572332 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48552 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 460941 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469343 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 550005 | AK033738.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 933140 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 276811 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 261336 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235886 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323108 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 233086 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 167894 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 144633 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 105570 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160505 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07191841 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02218014 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07191730 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07191867 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07191774 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02218133 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07191815 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160505 | - | - |
| Whole Mouse Genome G4122A | A_52_P311327 | AK006160.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P311327 | AK015597.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P311327 | AK076938.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122424 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160505 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02218014 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07191841 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07191815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08868484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08700667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08586057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27446007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08700683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08756521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08506877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08518830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08357052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08518834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08700692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08700701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27446011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08880147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27446001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08360235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08508128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08700664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08880146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08420610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27446014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08439994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08952456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08265970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08251991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08506871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27445894 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR1000021672.mm.1 | NR_045979.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000021672.mm.1 | NR_045979.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000021672.mm.1 | TC1000002866.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000021663.mm.1 | NR_045979.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000021663.mm.1 | NR_045979.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000021663.mm.1 | TC1000002866.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000021673.mm.1 | NR_045979.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000021673.mm.1 | NR_045979.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000021673.mm.1 | TC1000002866.mm.1 | MTA TC |

1700020G17Rik
Genomic Map