Assay Details
| Gene Symbol: Gm10865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045746.1 | 3 | 622 | 97 | ||
| GenBank mRNA | AK139534.1 | - | 3 | 623 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TRIO and F-actin binding protein | Triobp |
AI428493, EST478828, Tara, mKIAA1662 |
Anti-sense | Chr. 15 - 78947724 - 79005869 on NCBI Build GRCm38 | |
| Downstream | microRNA 6956 | Mir6956 |
Gm27880, mmu-mir-6956 |
6.8kb | Anti-sense | Chr. 15 - 79002223 - 79002284 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay targets the same location as MTA transcript cluster(s) TC1500001856.mm
This assay targets the same location as RTA transcript cluster(s) TC0700003481.rn
This assay targets the same location as RTA transcript cluster(s) TC0700003481.rn
Set Membership: |
|

Gm10865
Genomic Map